Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Flt-3 Ligand/FLT3L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB308500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB308500 500 μg
MAB308SP 25 μg
MAB308100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB308500 Supplier R&D Systems Supplier No. MAB308500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Flt-3 Ligand/FLT3L Monoclonal specifically detects Flt-3 Ligand/FLT3L in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Flt-3 Ligand
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 40406
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.02-0.06 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA17999
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Gene Symbols FLT3LG
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Flt-3 Ligand Thr27-Pro185 Accession # AAA17999
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2323
Test Specificity Detects human Flt-3 Ligand in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse Flt-3 Ligand.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.