Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Flt-3 Ligand/FLT3L Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $561.50
Specifications
Antigen | Flt-3 Ligand/FLT3L |
---|---|
Clone | 40406R |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB308RSP
![]() |
R&D Systems
MAB308RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB308R100
![]() |
R&D Systems
MAB308R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
Flt-3 Ligand/FLT3L Monoclonal specifically detects Flt-3 Ligand/FLT3L in Human samples. It is validated for Neutralization.Specifications
Flt-3 Ligand/FLT3L | |
Neutralization | |
Unconjugated | |
Mouse | |
Human | |
AAA17999 | |
2323 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
40406R | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Flt-3 Ligand, Thr27-Pro185, Accession # AAA17999 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title