Learn More
Description
Specifications
Specifications
| Assay Range | 12.5 pg/ml-200 pg/ml |
| Assay Sensitivity | 6.25 pg/ml |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| For Use With (Application) | ELISA |
| Host Species | Human |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
| Target | Folic acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
