Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Follistatin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6692SP
This item is not returnable.
View return policy
Description
Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for ELISA.Specifications
Follistatin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
follistatin, follistatin isoform FST317, FS, FSActivin-binding protein, FST | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Follistatin Gly30-Asp329 Accession # P19883 | |
25 μg | |
10468 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
ELISA | |
85943 | |
ELISA | |
P19883 | |
Mouse | |
Protein A or G purified from ascites | |
Primary | |
Detects human Follistatin in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction