Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Follistatin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB669500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB669500 500 μg
MAB669SP 25 μg
MAB669100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB669500 Supplier R&D Systems Supplier No. MAB669500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Follistatin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 85918
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P19883
Gene Alias follistatin, follistatin isoform FST317, FS, FSActivin-binding protein, FST
Gene Symbols FST
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Follistatin Gly30-Asp329 Accession # P19883
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10468
Test Specificity Detects human Follistatin in direct ELISAs and Western blots. It recognizes the 288 aa, 300 aa, and 315 aa isoforms of human Follistatin. In direct ELISAs and Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse Follistatin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.