Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Follistatin-like 1/FSTL1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1694SP
This item is not returnable.
View return policy
Description
Follistatin-like 1/FSTL1 Monoclonal specifically detects Follistatin-like 1/FSTL1 in Human samples. It is validated for Western Blot.Specifications
Follistatin-like 1/FSTL1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q12841 | |
FSTL1 | |
Mouse myeloma cell line NS0-derived recombinant human Follistatin-like 1/FSTL1 Met1-Ile308 Accession # Q12841 | |
25 μg | |
Primary | |
Detects human Follistatin-like 1/FSTL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse FSTL1 is observed. | |
Human | |
Purified |
Western Blot | |
229007 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Flik, FLJ50214, follistatin-like 1, Follistatin-like protein 1, Follistatin-related Protein, follistatin-related protein 1, FRP, FRPFLJ52277, FSL1, FSTL1, TSC-36 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11167 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction