Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Follistatin-related Gene Protein/FLRG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1288
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1288 500 μg
MAB1288SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1288 Supplier R&D Systems Supplier No. MAB1288
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Follistatin-related Gene Protein/FLRG/Fstl3 Monoclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Follistatin-related Gene Protein/FLRG/Fstl3
Applications ELISA
Classification Monoclonal
Clone 206714
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95633
Gene Alias FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3
Gene Symbols FSTL3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Follistatin-related Gene Protein/FLRG Met27-Val263 Accession # O95633
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272
Test Specificity Detects human Follistatin-related Gene Protein/FLRG in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant mouse FLRG is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.