Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Follistatin-related Gene Protein/FLRG Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1288
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1288 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1288 Supplier R&D Systems Supplier No. BAF1288
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Follistatin-related Gene Protein/FLRG/Fstl3 Polyclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Follistatin-related Gene Protein/FLRG/Fstl3
Applications ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. O95633
Gene Alias FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3
Gene Symbols FSTL3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Follistatin-related Gene Protein/FLRG Met27-Val263 Accession # O95633
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10272
Test Specificity Detects human Follistatin-related Gene Protein/FLRG in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant mouse FLRG is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.