Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FOLR1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5646T100UG
This item is not returnable.
View return policy
Description
FOLR1 Monoclonal specifically detects FOLR1 in Human samples. It is validated for Flow Cytometry.Specifications
FOLR1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P15328 | |
FOLR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human FOLR1 Arg25-Met233 Accession # P15328 | |
RUO | |
2348 | |
Human | |
IgG1 |
Flow Cytometry | |
548908 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
Mouse | |
100 μg | |
Primary | |
Detects human FOLR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FOLR2, 3 or 4 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction