Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FOLR1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems BAF5646
This item is not returnable.
View return policy
Description
FOLR1 Polyclonal specifically detects FOLR1 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
FOLR1 | |
Polyclonal | |
Biotin | |
P15328 | |
FOLR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human FOLR1 Arg25-Met233 Accession # P15328 | |
50 μg | |
Primary | |
Detects human FOLR1 in ELISAs and Western blots. In sandwich ELISAs, less than 0.2% cross-reactivity with recombinant human (rh) FOLR2, rhFOLR3, rhFOLR4 and recombinant mouse FOLR1 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
Goat | |
Affinity Purified | |
RUO | |
2348 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction