Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FOLR1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB5646P
This item is not returnable.
View return policy
Description
FOLR1 Monoclonal specifically detects FOLR1 in Human samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
FOLR1 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2348 | |
Human | |
Purified |
Flow Cytometry, Immunoassay | |
548908 | |
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells | |
P15328 | |
FOLR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human FOLR1 Arg25-Met233 Accession # P15328 | |
100 Tests | |
Primary | |
Detects human FOLR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FOLR2, 3 or 4 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction