Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FOLR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5697SP
This item is not returnable.
View return policy
Description
FOLR2 Monoclonal specifically detects FOLR2 in Human samples. It is validated for Immunocytochemistry.Specifications
FOLR2 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P14207 | |
FOLR2 | |
Chinese hamster ovary cell line CHO-derived recombinant human FOLR2 Gln22-His228 Accession # P14207 | |
25 μg | |
Primary | |
Detects human FOLR2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human FOLR1, 3,or 4 is observed. | |
Human | |
Purified |
Immunocytochemistry | |
583121 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2350 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction