Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FOLR3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5319
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5319 Supplier R&D Systems Supplier No. AF5319
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FOLR3 Polyclonal specifically detects FOLR3 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen FOLR3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. P41439
Gene Alias Folate receptor 3, folate receptor 3 (gamma), folate receptor gamma, FR-G, FR-gamma, gamma-hFR
Gene Symbols FOLR3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human FOLR3 long isoform Ala20-Ser243 Accession # P41439
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2352
Test Specificity Detects human FOLR3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FOLR2 is observed and less than 1% cross-reactivity with rhFOLR1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.