Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FOLR4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB6328SP
This item is not returnable.
View return policy
Description
FOLR4 Monoclonal specifically detects FOLR4 in Human samples. It is validated for Immunohistochemistry.Specifications
FOLR4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
folate receptor 4 (delta) homolog (mouse), Folbp3, FR-delta, LOC390243, probable folate receptor delta | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
390243 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Immunohistochemistry | |
821221 | |
Immunohistochemistry 8-25 ug/mL | |
A6ND01 | |
FOLR4 | |
E. coli-derived recombinant human FOLR4 Glu134-Trp210 Accession # A6ND01 | |
25 μg | |
Primary | |
Detects human FOLR4 in direct ELISAs. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction