Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FosB/G0S3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2214
This item is not returnable.
View return policy
Description
FosB/G0S3 Polyclonal specifically detects FosB/G0S3 in Human samples. It is validated for Western Blot.Specifications
FosB/G0S3 | |
Polyclonal | |
Biotin | |
P53539 | |
FOSB | |
E. coli-derived recombinant human FosB/G0S3 Met1-Leu338 Accession # P53539 | |
50 μg | |
Primary | |
Detects human FosB/G0S3 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
activator protein 1, AP-1, DKFZp686C0818, FBJ murine osteosarcoma viral oncogene homolog B, G0S3, G0S3G0/G1 switch regulatory protein 3, GOS3, GOSB, MGC42291, oncogene FOS-B, protein fosB | |
Goat | |
Affinity Purified | |
RUO | |
2354 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction