Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxC2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5044
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5044 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5044 Supplier R&D Systems Supplier No. MAB5044
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FoxC2 Monoclonal specifically detects FoxC2 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FoxC2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 665819
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99958
Gene Alias Fkh14, FKHL14LD, forkhead box C2 (MFH-1, mesenchyme forkhead 1), forkhead, Drosophila, homolog-like 14, Forkhead-related protein FKHL14, LD, Mesenchyme fork head protein 1, mesenchyme forkhead 1, MFH1, MFH-1, MFH-1 protein, MFH1forkhead box protein C2, Transcription factor FKH-14
Gene Symbols FOXC2
Host Species Mouse
Immunogen E. coli-derived recombinant human FoxC2 Gly415-Tyr501 Accession # Q99958
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2303
Test Specificity Detects human FoxC2 in direct ELISAs and Western blots. Approximately 25% cross-reactivity with recombinant human (rh) FoxA1 (aa 253-472) is observed in Western blots but not in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with rhFoxB1 (aa 191-324), D3 (aa 1-140), F1 (aa 129-354), J1 (aa 306-421), K1 (aa 493-670), M1 (aa 642-763), P1 (aa 231-420), P2 (aa 269-460), P3 (aa 179-279), or P4 (aa 229-410) is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.