Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxD3 aa 1-140 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2819
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2819 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2819 Supplier R&D Systems Supplier No. AF2819
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FoxD3 Polyclonal specifically detects FoxD3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FoxD3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9UJU5
Gene Alias AIS1, CWH3, forkhead box D3, forkhead box protein D3, Genesis, HFH2, HNF3/FH transcription factor genesis
Gene Symbols FOXD3
Host Species Sheep
Immunogen E. coli-derived recombinant human FoxD3 Met1-Val140 Accession # Q9UJU5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27022
Test Specificity Detects human FoxD3 (aa 1-140) in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxP3, rhFoxP1, and rhFoxO3A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.