Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FOXL2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $467.50

Specifications

Antigen FOXL2
Clone 1019104
Dilution Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10396SP
GREENER_CHOICE
View Documents
R&D Systems
MAB10396SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB10396100
GREENER_CHOICE
View Documents
R&D Systems
MAB10396100
100 μg
Each for $467.50
Only null left
Add to Cart
 
Description

Description

FOXL2 Monoclonal specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

FOXL2
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
BPES, BPES1, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, PINTO, POF3
Synthetic peptide containing human FOXL2, Accession # P58012
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1019104
Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
Human
P58012
668
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.