Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxM1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3975SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3975SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3975SP Supplier R&D Systems Supplier No. AF3975SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FoxM1 Polyclonal specifically detects FoxM1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FoxM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q08050
Gene Alias FKHL16, FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH-11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, Trident, WIN, Winged-helix factor from INS-1 cells
Gene Symbols FOXM1
Host Species Sheep
Immunogen E. coli-derived recombinant human FoxM1 Thr642-Gln763 Accession # Q08050
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2305
Test Specificity Detects human FoxM1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FoxB2 and rhFoxJ1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.