Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxM1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3975
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3975 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3975 Supplier R&D Systems Supplier No. BAF3975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FoxM1 Polyclonal specifically detects FoxM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FoxM1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q08050
Gene Alias FKHL16, FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH-11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, Trident, WIN, Winged-helix factor from INS-1 cells
Gene Symbols FOXM1
Host Species Sheep
Immunogen E. coli-derived recombinant human FoxM1 Thr642-Gln763 Accession # Q08050
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2305
Test Specificity Detects human FoxM1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) FoxJ1 is observed and less than 1% cross-reactivity with rhFoxB2, rhFoxC2, rhFoxD3, rhFoxF1, rhFoxH1, rhFoxO3A, rhFoxP1, and rhFoxP3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.