Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FoxO1/FKHR Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5939SP
This item is not returnable.
View return policy
Description
FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot, Simple Western.Specifications
FoxO1/FKHR | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FKH1, FKHR, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
Sheep | |
Affinity Purified | |
RUO | |
2308 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
Q12778 | |
FOXO1 | |
E. coli-derived recombinant human FoxO1/FKHR Ala353-Gly655 Accession # Q12778 | |
25 μg | |
Primary | |
Detects human FoxO1/FKHR in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction