Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FoxP3 Buffer Set; BD

Catalog No. BDB560098 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560098 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560098 Supplier BD Biosciences Supplier No. 560098
Add to Cart
Add to Cart

Human FoxP3 Buffer Set is optimized for use with the FoxP3 mAb clone 259D/C7. It is intented for the fixation and permeabilization of PBMCs (orlysed whole blood) for intracellular staining of Human FoxP3 and surface staining of the appropriate CD markers. See recommended assay procedure for the Human FoxP3 Buffer Set and the FoxP3 conjugates for the proper use and handling of these products.

Kit components:
Human FoxP3 Buffer A (10 X) 100tests
Human FoxP3 Buffer bA (50 X) 100tests

Applications:

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.