Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FPR1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3744T100UG
This item is not returnable.
View return policy
Description
FPR1 Monoclonal specifically detects FPR1 in Human samples. It is validated for Flow Cytometry.Specifications
FPR1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_002020 | |
FPR1 | |
HEK293 human embryonic kidney cell line transfected with human FPR1 Met1-Lys350 Accession # NP_002020 | |
RUO | |
2357 | |
Human | |
IgG2a |
Flow Cytometry | |
350418 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
fMet-Leu-Phe receptor, fMLF-R, FMLP, fMLP receptor, formyl peptide receptor 1, FPRfMet-Leu-Phe receptor, N-formyl peptide receptor, N-formylpeptide chemoattractant receptor | |
Mouse | |
100 μg | |
Primary | |
Detects human FPR1. Stains human FPR1-transfected cells but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction