Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FPRL1/FPR2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3479R100UG
This item is not returnable.
View return policy
Description
FPRL1/FPR2 Monoclonal specifically detects FPRL1/FPR2 in Human samples. It is validated for Flow Cytometry.Specifications
FPRL1/FPR2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P25090 | |
FPR2 | |
NS0 mouse myeloma cell line transfected with human FPRL1 Met1-Met351 Accession # P25090 | |
RUO | |
2358 | |
Reconstitution default not known | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
304405 | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ALXR, FMLP-R-I, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4 Receptor, LXA4RFMLP-R-I, N-formyl peptide receptor 2, RFP | |
Mouse | |
100 μg | |
Primary | |
Stains FPRL1 transfectants but not irrelevant transfectants. It also stains monocytes and granulocytes. | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction