Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FPRL1/FPR2 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3479C
This item is not returnable.
View return policy
Description
FPRL1/FPR2 Monoclonal specifically detects FPRL1/FPR2 in Human samples. It is validated for Flow Cytometry.Specifications
FPRL1/FPR2 | |
Monoclonal | |
PerCP | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ALXR, FMLP-R-I, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4 Receptor, LXA4RFMLP-R-I, N-formyl peptide receptor 2, RFP | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2358 | |
Human | |
Purified |
Flow Cytometry | |
304405 | |
Flow Cytometry 10 uL/10^6 cells | |
P25090 | |
FPR2 | |
NS0 mouse myeloma cell line transfected with human FPRL1/FPR2 Met1-Met351 Accession # P25090 | |
100 Tests | |
Primary | |
Stains FPRL1/FPR2 transfectants but not irrelevant transfectants. It also stains monocytes and granulocytes. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction