Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FPRL2/FPR3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3896SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3896SP 25 μg
MAB3896 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3896SP Supplier R&D Systems Supplier No. MAB3896SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FPRL2/FPR3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 374822
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P25089
Gene Alias FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPR3, FPRH1, FPRH1FPRH2, FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Gene Symbols FPR3
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FPRL2 Met1-Met352 Accession # P25089
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2359
Test Specificity Detects human FPRL2. Stains human FPRL2 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.