Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FPRL2/FPR3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3896A
This item is not returnable.
View return policy
Description
FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry.Specifications
FPRL2/FPR3 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPR3, FPRH1, FPRH1FPRH2, FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2359 | |
Human | |
Purified |
Flow Cytometry | |
374822 | |
Flow Cytometry 10 uL/10^6 cells | |
P25089 | |
FPR3 | |
HEK293 human embryonic kidney cell line transfected with human FPRL2 Met1-Met352 Accession # P25089 | |
100 Tests | |
Primary | |
Stains human FPRL2 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction