Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Frk Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3766SP
This item is not returnable.
View return policy
Description
Frk Monoclonal specifically detects Frk in Human samples. It is validated for Immunohistochemistry.Specifications
Frk | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P42685 | |
FRK | |
E. coli-derived recombinant human Frk Met1-Glu107 Accession # P42685 | |
25 μg | |
Primary | |
Detects human Frk in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
393812 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, Gtk, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5, PTK5 protein tyrosine kinase 5, Rak, RAKPTK5, tyrosine-protein kinase FRK | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2444 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction