Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FRS2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC40691S100UG
This item is not returnable.
View return policy
Description
FRS2 Monoclonal specifically detects FRS2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FRS2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8WU20 | |
FRS2 | |
E. coli-derived recombinant human FRS2 Asn121-Asn449 Accession # Q8WU20 | |
RUO | |
10818 | |
Human | |
IgG2b |
Flow Cytometry | |
462902 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1 | |
Mouse | |
100 μg | |
Primary | |
Detects human FRS2 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction