Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FSH beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB43102100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB43102100 100 μg
MAB43102SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB43102100 Supplier R&D Systems Supplier No. MAB43102100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

FSH beta Monoclonal specifically detects FSH beta in Human samples. It is validated for Western Blot.

Specifications

Antigen FSH beta
Applications Western Blot
Classification Monoclonal
Clone 961417
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P01225
Gene Alias follicle stimulating hormone, beta polypeptide, Follicle-stimulating hormone beta subunit, Follitropin, Follitropin beta chain, follitropin subunit beta, follitropin, beta chain, FSHB, FSH-B, FSH-beta
Host Species Rat
Immunogen E.coli-derived recombinant human FSH beta Asn19-Glu129 Accession # P01225
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Test Specificity Detects human FSH beta in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.