Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FUBI/MNSF beta Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | FUBI/MNSF beta/FAU |
---|---|
Clone | 861504 |
Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Description
FUBI/MNSF beta/FAU Monoclonal specifically detects FUBI/MNSF beta/FAU in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FUBI/MNSF beta/FAU | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
40S ribosomal protein S30, asr1, FAU, FAU1, FAU-encoded ubiquitin-like protein, FBR-MuSV-associated ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed(fox derived), FLJ22986, Fub1, Fubi, MNSFbeta, monoclonal nonspecific suppressor factor beta, ribosomal protein S30, RPS30, S30, ubiquitin-like protein fubi and ribosomal protein S30, ubiquitin-like-S30 fusion protein | |
FAU | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
861504 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rat | |
Human | |
P35544 | |
2197 | |
E.coli-derived recombinant human FUBI/MNSF beta Met1-Gly74 Accession # P35544 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FUBI in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title