Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fucosyltransferase 11/FUT11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5964SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5964SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5964SP Supplier R&D Systems Supplier No. AF5964SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Fucosyltransferase 11/FUT11 Polyclonal specifically detects Fucosyltransferase 11/FUT11 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Fucosyltransferase 11/FUT11
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH36037
Gene Alias alpha-(1,3)-fucosyltransferase 11, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 11, fucosyltransferase 11 (alpha (1,3) fucosyltransferase), Fucosyltransferase XI, Fuc-TXI, FucT-XI, FUT11, Galactoside 3-L-fucosyltransferase 11, MGC119338, MGC119339, MGC33202
Gene Symbols FUT11
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 11/FUT11 Gly25-Leu492 Accession # AAH36037
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170384
Test Specificity Detects human FUT11 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FUT3, rhFUT5, and rhFUT8 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.