Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fucosyltransferase 7/FUT7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB64091SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB64091SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB64091SP Supplier R&D Systems Supplier No. MAB64091SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fucosyltransferase 7/FUT7 Monoclonal specifically detects Fucosyltransferase 7/FUT7 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Fucosyltransferase 7/FUT7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 795116
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q11130
Gene Alias alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, FTVII, Fucosyltransferase 7, fucosyltransferase 7 (alpha (1,3) fucosyltransferase), Fucosyltransferase VII, Fuc-TVII, FucT-VII, FUT7, Galactoside 3-L-fucosyltransferase, Selectin ligand synthase, selectin-ligand synthase
Gene Symbols FUT7
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 7/FUT7 Ala36-Ala342 Accession # Q11130
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2529
Test Specificity Detects human Fucosyltransferase 7/FUT7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Fucosyltransferase 5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.