Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fucosyltransferase 7/FUT7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB64091
This item is not returnable.
View return policy
Description
Fucosyltransferase 7/FUT7 Monoclonal specifically detects Fucosyltransferase 7/FUT7 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Fucosyltransferase 7/FUT7 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q11130 | |
FUT7 | |
Chinese hamster ovary cell line CHO-derived recombinant human Fucosyltransferase 7/FUT7 Ala36-Ala342 Accession # Q11130 | |
100 μg | |
Primary | |
Detects human Fucosyltransferase 7/FUT7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Fucosyltransferase 5 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
795116 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, FTVII, Fucosyltransferase 7, fucosyltransferase 7 (alpha (1,3) fucosyltransferase), Fucosyltransferase VII, Fuc-TVII, FucT-VII, FUT7, Galactoside 3-L-fucosyltransferase, Selectin ligand synthase, selectin-ligand synthase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2529 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction