Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Furin Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1503U100UG
This item is not returnable.
View return policy
Description
Furin Monoclonal specifically detects Furin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Furin | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_002560 | |
FURIN | |
Mouse myeloma cell line NS0-derived recombinant human Furin Asp108-Glu715 Accession # NP_002560 | |
RUO | |
5045 | |
Human | |
IgG2b |
Flow Cytometry | |
222722 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
Mouse | |
100 μg | |
Primary | |
Detects human Furin in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction