Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Furin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1503
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1503 100 μg
AF1503SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1503 Supplier R&D Systems Supplier No. AF1503
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

Furin Polyclonal specifically detects Furin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Furin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P09958
Gene Alias Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FUR, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACE, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PC, PCSK3, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Gene Symbols FURIN
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Furin Asp108-Glu715 Accession # P09958
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5045
Test Specificity Detects human Furin in direct ELISAs and Western blots. In direct ELISAs, less than 40% cross-reactivity with recombinant mouse Furin is observed, and less than 1% cross-reactivity with recombinant human (rh) PC-1 and rhPC-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.