Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FXR/NR1H4 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. PPA9033A00 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPA9033A00 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPA9033A00 Supplier R&D Systems Supplier No. PPA9033A00

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

FXR/NR1H4 Monoclonal specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Direct ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen FXR/NR1H4
Applications Direct ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A9033A
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 with 0.1% Sodium Azide
Gene Alias BAR, bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, NR1H4, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14, RIP14RXR-interacting protein 14
Gene Symbols NR1H4
Host Species Mouse
Immunogen Recombinant human FXR/NR1H4 aa 2-126
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9971
Test Specificity This antibody specifically recognizes human FXR and cross-reacts with rat and mouse FXR. Not yet tested in other species.
Target Species Human, Mouse
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.