Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FXYD5/Dysadherin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF8300
This item is not returnable.
View return policy
Description
FXYD5/Dysadherin Polyclonal specifically detects FXYD5/Dysadherin in Human samples. It is validated for Immunohistochemistry.Specifications
FXYD5/Dysadherin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DYSAD, Dysadherin, EF-8, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
Goat | |
Affinity Purified | |
RUO | |
53827 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
Q96DB9 | |
FXYD5 | |
E. coli-derived recombinant human FXYD5/Dysadherin Gln22-Asp133 Accession # Q96DB9 | |
100 μg | |
Primary | |
Detects human FXYD5/Dysadherin in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction