Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FYB/ADAP/SLAP130 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC38571U100UG
This item is not returnable.
View return policy
Description
FYB/ADAP/SLAP130 Monoclonal specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| FYB/ADAP/SLAP130 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15117 | |
| FYB | |
| E. coli-derived recombinant human FYB/ADAP/SLAP130 Ala2-Gly274 Accession # O15117 | |
| RUO | |
| 2533 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 460107 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FYB/ADAP/SLAP130 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction