Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FYB/ADAP/SLAP130 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC38571T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC38571T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC38571T100 Supplier R&D Systems Supplier No. IC38571T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FYB/ADAP/SLAP130 Monoclonal specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen FYB/ADAP/SLAP130
Applications Flow Cytometry
Classification Monoclonal
Clone 460107
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O15117
Gene Alias ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein
Gene Symbols FYB
Host Species Mouse
Immunogen E. coli-derived recombinant human FYB/ADAP/SLAP130 Ala2-Gly274 Accession # O15117
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2533
Test Specificity Detects human FYB/ADAP/SLAP130 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.