Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FYB/ADAP/SLAP130 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3857
This item is not returnable.
View return policy
Description
FYB/ADAP/SLAP130 Polyclonal specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Western Blot.Specifications
FYB/ADAP/SLAP130 | |
Polyclonal | |
Unconjugated | |
O15117 | |
FYB | |
E. coli-derived recombinant human FYB/ADAP/SLAP130 Ala2-Gly274 Accession # O15117 | |
100 μg | |
Primary | |
Detects human FYB/ADAP/SLAP130 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
Goat | |
Affinity Purified | |
RUO | |
2533 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction