Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human G-CSF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF214SP
This item is not returnable.
View return policy
Description
G-CSF Polyclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, Neutralization.Specifications
G-CSF | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
Goat | |
Affinity Purified | |
RUO | |
1440 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 8-48 ng/mL | |
NP_757373 | |
CSF3 | |
E. coli-derived recombinant human G-CSF Thr31-Pro204 Accession # NP_757373 | |
25 μg | |
Primary | |
Detects human G-CSF in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 30% cross-reactivity with recombinant mouse G-CSF and recombinant canine G-CSF is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction