Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human G-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB214NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB214NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB214NA Supplier R&D Systems Supplier No. AB214NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

G-CSF Polyclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen G-CSF
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.1-0.3 ug/mL
Gene Accession No. NP_757373
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Gene Symbols CSF3
Host Species Goat
Immunogen E. coli-derived recombinant human G-CSF Thr31-Pro204 Accession # NP_757373
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Test Specificity Detects human G-CSF in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse G-CSF is observed.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.