Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human G-CSF Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF214
This item is not returnable.
View return policy
Description
G-CSF Polyclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
G-CSF | |
Polyclonal | |
Biotin | |
NP_757373 | |
CSF3 | |
E. coli-derived recombinant human G-CSF Thr31-Pro204 Accession # NP_757373 | |
50 μg | |
Primary | |
Detects human G-CSF in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant mouse G-CSF and recombinant human G-CSF R is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
Goat | |
Affinity Purified | |
RUO | |
1440 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction