Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human G-CSFR/CD114 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3811U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3811U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3811U100 Supplier R&D Systems Supplier No. FAB3811U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

G-CSFR/CD114 Monoclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen G-CSFR/CD114
Applications Flow Cytometry
Classification Monoclonal
Clone 38643
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q99062
Gene Alias CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Gene Symbols CSF3R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114 Glu25-Pro621 Accession # Q99062
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Test Specificity Detects human G-CSF R/CD114 in ELISAs and Western blots. In sandwich Immunoassays, no cross-reactivity with recombinant human (rh) G-CSF, rhGM-CSF R beta , rhGM-CSF R, or recombinant mouse G-CSF is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.