Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human G-CSFR/CD114 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3811V100UG
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Monoclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for Flow Cytometry.Specifications
G-CSFR/CD114 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q99062 | |
CSF3R | |
Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114 Glu25-Pro621 Accession # Q99062 | |
RUO | |
1441 | |
Human | |
IgG1 |
Flow Cytometry | |
38643 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
Mouse | |
100 μg | |
Primary | |
Detects human G-CSF R/CD114 in ELISAs and Western blots. In sandwich Immunoassays, no cross-reactivity with recombinant human (rh) G-CSF, rhGM-CSF R beta , rhGM-CSF R, or recombinant mouse G-CSF is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction