Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human G-CSFR/CD114 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF381PB
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF381PB 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF381PB Supplier R&D Systems Supplier No. AF381PB
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

G-CSFR/CD114 Polyclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot.

Specifications

Antigen G-CSFR/CD114
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q99062
Gene Alias CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Gene Symbols CSF3R
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114 Glu25-Pro621 Accession # Q99062
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Test Specificity Detects human G-CSF R/CD114 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) G-CSF and rhGM-CSF R alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.