Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human G6PD Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $484.00
Specifications
Antigen | Glucose 6 Phosphate Dehydrogenase |
---|---|
Clone | 1067503 |
Dilution | Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL |
Applications | Western Blot, Simple Western, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11467SP
![]() |
R&D Systems
MAB11467SP |
25 μg |
Each for $208.00
|
|
|||||
MAB11467100
![]() |
R&D Systems
MAB11467100 |
100 μg |
Each for $484.00
|
|
|||||
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Simple Western, ImmunohistochemistrySpecifications
Glucose 6 Phosphate Dehydrogenase | |
Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
G6PD, G6PD1EC 1.1.1.49, G6PDH, glucose-6-phosphate dehydrogenase | |
E. coli-derived human G6PD, Ala2-Leu515, Accession # P11413 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1067503 | |
Western Blot, Simple Western, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P11413 | |
2539 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title