Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Gab2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4877SP
This item is not returnable.
View return policy
Description
GAB2 Polyclonal specifically detects GAB2 in Human samples. It is validated for Western Blot.Specifications
GAB2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GRB2-associated binding protein 2, GRB2-associated-binding protein 2, Growth factor receptor bound protein 2-associated protein 2, KIAA0571GRB2-associated binder 2, p100, pp100 | |
Sheep | |
Affinity Purified | |
RUO | |
9846 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.2 ug/mL | |
Q9UQC2 | |
GAB2 | |
E. coli-derived recombinant human Gab2 Gln82-Phe292 Accession # Q9UQC2 | |
25 μg | |
Primary | |
Detects endogenous human Gab2 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction