Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GAD1/GAD67 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2086
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2086 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2086 Supplier R&D Systems Supplier No. BAF2086
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GAD1/GAD67 Polyclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot.

Specifications

Antigen GAD1/GAD67
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q99259
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, GAD67, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Gene Symbols GAD1
Host Species Goat
Immunogen E. coli-derived recombinant human GAD1/GAD67 Ala2-Asn97 Accession # Q99259
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2571
Test Specificity Detects human GAD1/GAD67 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human GAD2/GAD65 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.